siRNA Details
| Gene Symbol: Bloc1s4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133724.3 | NP_598485.1 | 1 | 774 | ||
| GenBank mRNA | AK011795.1 | - | 1 | 746 | ||
| BC019169.1 | - | 1 | 764 | 5002867 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bloc1s4| Gene Symbol: Bloc1s4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133724.3 | NP_598485.1 | 1 | 774 | ||
| GenBank mRNA | AK011795.1 | - | 1 | 746 | ||
| BC019169.1 | - | 1 | 764 | 5002867 | ||