siRNA Details
| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054103.1 | NP_473444.1 | 4, 5 | 798 | ||
| GenBank mRNA | AK014819.1 | - | 4, 5 | 798 | ||
| AM056057.1 | - | 1, 2 | 66 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk33| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054103.1 | NP_473444.1 | 4, 5 | 798 | ||
| GenBank mRNA | AK014819.1 | - | 4, 5 | 798 | ||
| AM056057.1 | - | 1, 2 | 66 | |||