siRNA Details
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080446.2 | NP_536694.2 | 10, 11 | 2562 | ||
GenBank mRNA | AB048542.1 | - | 10, 11 | 2487 | ||
AK131122.1 | - | 10, 11 | 2584 | |||
BC068140.1 | - | 10, 11 | 2490 | 6843107 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
