siRNA Details
| Gene Symbol: Mrps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080456.1 | NP_536704.1 | 3 | 320 | ||
| GenBank mRNA | AB049943.1 | - | 3 | 335 | ||
| AK165590.1 | - | 3 | 337 | |||
| AK166374.1 | - | 3 | 335 | |||
| AK167304.1 | - | 3 | 335 | |||
| AK167316.1 | - | 3 | 335 | |||
| AY061856.1 | - | 3 | 335 | |||
| BC027271.1 | - | 3 | 320 | 3156971 | ||

Mrps6
Genomic Map