siRNA Details
| Gene Symbol: Strc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080459.2 | NP_536707.2 | 6, 7 | 2494 | ||
| GenBank mRNA | AF375593.1 | - | 6, 7 | 2494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strc| Gene Symbol: Strc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080459.2 | NP_536707.2 | 6, 7 | 2494 | ||
| GenBank mRNA | AF375593.1 | - | 6, 7 | 2494 | ||