siRNA Details
| Gene Symbol: Man2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172903.4 | NP_766491.2 | 14, 15 | 2444 | ||
| GenBank mRNA | AK029913.1 | - | 14, 15 | 2255 | ||
| AK044028.1 | - | 13, 14 | 3182 | |||
| AK044561.1 | - | 3, 4 | 712 | |||
| AK161799.1 | - | 13, 14 | 2005 | |||
| BC055006.1 | - | 3, 4 | 500 | 6491507 | ||
| DQ631806.1 | - | 12, 13 | 1959 | |||

Man2a2
Genomic Map