siRNA Details
| Gene Symbol: Zfp358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080461.2 | NP_536709.2 | 2 | 316 | ||
| GenBank mRNA | AF397208.1 | - | 2 | 282 | ||
| AK155446.1 | - | 2 | 318 | |||
| BC055855.1 | - | 1 | 517 | |||
| BY749799.1 | - | 2 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp358| Gene Symbol: Zfp358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080461.2 | NP_536709.2 | 2 | 316 | ||
| GenBank mRNA | AF397208.1 | - | 2 | 282 | ||
| AK155446.1 | - | 2 | 318 | |||
| BC055855.1 | - | 1 | 517 | |||
| BY749799.1 | - | 2 | 284 | |||