siRNA Details
| Gene Symbol: Smc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080470.1 | NP_536718.1 | 6 | 1048 | ||
| GenBank mRNA | AF303827.1 | - | 6 | 1048 | ||
| BC129800.1 | - | 6 | 1039 | 40048794 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc1b| Gene Symbol: Smc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080470.1 | NP_536718.1 | 6 | 1048 | ||
| GenBank mRNA | AF303827.1 | - | 6 | 1048 | ||
| BC129800.1 | - | 6 | 1039 | 40048794 | ||