siRNA Details
| Gene Symbol: Plxnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019587.2 | NP_062533.2 | 12 | 2353 | ||
| GenBank mRNA | AK122467.1 | - | 11 | 1279 | ||
| AK138168.1 | - | 10 | 1180 | |||
| AK138631.1 | - | 12 | 1428 | |||
| AK140240.1 | - | 12 | 2357 | |||

Plxnb3
Genomic Map