siRNA Details
| Gene Symbol: Sec63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153055.3 | NP_694695.3 | 1, 2 | 397 | ||
| GenBank mRNA | AK036629.1 | - | 1, 2 | 92 | ||
| AK049737.1 | - | 1, 2 | 88 | |||
| AY024346.1 | - | 1, 2 | 194 | |||
| AY238936.1 | - | 1, 2 | 167 | |||
| BC019366.1 | - | 1, 2 | 161 | 5066908 | ||
| BC031846.1 | - | 1, 2 | 161 | 5066822 | ||
| BC059816.1 | - | 1, 2 | 397 | |||
| BC080810.1 | - | 1, 2 | 278 | |||

Sec63
Genomic Map