siRNA Details
Gene Symbol: Gpr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199058.1 | NP_951013.1 | 1 | 391 | ||
GenBank mRNA | AK139367.1 | - | 2 | 570 | ||
AY064488.1 | - | 1 | 391 | |||
BC111861.1 | - | 1 | 380 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
