siRNA Details
| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163776.1 | NP_001157248.1 | 12 | 1491 | ||
| NM_080727.2 | NP_542765.2 | 11 | 1351 | |||
| GenBank mRNA | AJ300738.1 | - | 11 | 1351 | ||
| AK158203.1 | - | 12 | 1494 | |||
| BC112375.1 | - | 11 | 1348 | 40057032 | ||

Tmprss3
Genomic Map