siRNA Details
| Gene Symbol: Myh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080728.2 | NP_542766.1 | 19 | 2113 | ||
| GenBank mRNA | AK040973.1 | - | 19 | 2115 | ||
| AK137508.1 | - | 1 | 891 | |||
| AK147031.1 | - | 19 | 2114 | |||
| AK157742.1 | - | 19 | 2114 | |||
| AK168677.1 | - | 3 | 144 | |||
| AK169198.1 | - | 19 | 2113 | |||
| AK169371.1 | - | 19 | 2113 | |||
| AY056464.1 | - | 19 | 2113 | |||
| BC121789.1 | - | 18 | 2069 | 40099448 | ||
| BC158018.1 | - | 2190 | 9007324 | |||
| BC158070.1 | - | 19 | 2368 | 9007368 | ||

Myh7
Genomic Map