siRNA Details
| Gene Symbol: Ttbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024856.2 | NP_001020027.1 | 8 | 1093 | ||
| NM_001024857.2 | NP_001020028.1 | 7 | 1019 | |||
| NM_080788.3 | NP_542966.2 | 8 | 847 | |||
| GenBank mRNA | AK046978.1 | - | 8 | 1066 | ||
| AK053820.1 | - | 8 | 1082 | |||
| AK077011.1 | - | 8 | 868 | |||
| AK137024.1 | - | 7 | 1008 | |||
| AK148269.1 | - | 7 | 1023 | |||

Ttbk2
Genomic Map