siRNA Details
| Gene Symbol: Caln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021371.2 | NP_067346.1 | 5 | 1163 | ||
| NM_181045.1 | NP_851388.1 | 6 | 863 | |||
| GenBank mRNA | AB041539.1 | - | 5 | 1154 | ||
| AF282251.1 | - | 6 | 845 | |||
| AK035865.1 | - | 6 | 832 | |||
| AK081011.1 | - | 5 | 597 | |||
| AK158641.1 | - | 5 | 525 | |||
| AK158892.1 | - | 5 | 1135 | |||
| AK163648.1 | - | 5 | 626 | |||
| BC057129.1 | - | 5 | 1142 | 6811387 | ||
| BC058963.1 | - | 5 | 1142 | 6811387 | ||

Caln1
Genomic Map