siRNA Details
| Gene Symbol: Zcchc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080855.2 | NP_543131.1 | 12, 13 | 2855 | ||
| GenBank mRNA | AB030244.1 | - | 12, 13 | 2866 | ||
| BC094454.1 | - | 1, 2 | 117 | |||
| BC132449.1 | - | 12, 13 | 2853 | 40130726 | ||
| BC137853.1 | - | 12, 13 | 2853 | 8860872 | ||

Zcchc14
Genomic Map