siRNA Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170748.1 | NP_001164219.1 | 8 | 1402 | ||
| NM_080856.4 | NP_543132.3 | 8 | 1402 | |||
| GenBank mRNA | AF403042.1 | - | 3 | 819 | ||
| AK052884.1 | - | 8 | 1404 | |||
| AK164800.1 | - | 2 | 465 | |||
| BC145002.1 | - | 3 | 788 | |||

Asb14
Genomic Map