siRNA Details
| Gene Symbol: Asb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267724.1 | NP_001254653.1 | 2 | 226 | ||
| NM_178283.4 | NP_840068.1 | 4 | 565 | |||
| GenBank mRNA | AF403041.1 | - | 4 | 565 | ||
| AK078289.1 | - | 4 | 521 | |||
| AK133892.1 | - | 4 | 491 | |||
| BC018240.1 | - | 4 | 519 | 4239996 | ||
| BG080214.2 | - | 2 | 160 | |||

Asb13
Genomic Map