siRNA Details
| Gene Symbol: Slc2a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130451.3 | NP_569718.1 | 3 | 1502 | ||
| GenBank mRNA | AK052156.1 | - | 3 | 1469 | ||
| AY029579.1 | - | 3 | 1425 | |||
| BC137917.1 | - | 3 | 1420 | 8860936 | ||
| BC137920.1 | - | 3 | 1420 | 8860939 | ||

Slc2a10
Genomic Map