siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 2 | 800 | ||
| GenBank mRNA | AB038773.1 | - | 2 | 134 | ||
| AB059999.1 | - | 2 | 314 | |||
| AK029060.1 | - | 2 | 796 | |||
| AK083513.1 | - | 2 | 796 | |||
| BC023769.1 | - | 1 | 92 | |||
| BC053086.1 | - | 2 | 614 | |||
| BC059000.1 | - | 2 | 774 | |||
| BC060070.1 | - | 2 | 614 | |||
| BC066176.1 | - | 2 | 523 | |||

Recql5
Genomic Map