siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 18, 19 | 3422 | ||
| GenBank mRNA | AB059999.1 | - | 18, 19 | 2936 | ||
| AK049269.1 | - | 13, 14 | 1822 | |||
| AK050965.1 | - | 16, 17 | 2151 | |||
| BC019613.1 | - | 4, 5 | 1208 | |||
| BC023769.1 | - | 17, 18 | 2714 | |||
| BC053086.1 | - | 18, 19 | 3675 | |||
| BC059000.1 | - | 18, 19 | 3395 | |||
| BC060070.1 | - | 18, 19 | 3675 | |||
| BC066176.1 | - | 18, 19 | 3145 | |||

Recql5
Genomic Map