siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 3 | 1050 | ||
| GenBank mRNA | AB059999.1 | - | 3 | 564 | ||
| AK083513.1 | - | 3 | 1046 | |||
| BC023769.1 | - | 2 | 342 | |||
| BC053086.1 | - | 3 | 864 | |||
| BC059000.1 | - | 3 | 1024 | |||
| BC060070.1 | - | 3 | 864 | |||
| BC066176.1 | - | 3 | 773 | |||

Recql5
Genomic Map