siRNA Details
| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130455.2 | NP_569722.1 | 3 | 1954 | ||
| GenBank mRNA | AF373861.1 | - | 1 | 703 | ||
| AF396649.1 | - | 3 | 1963 | |||
| AY062435.1 | - | 3 | 1954 | |||
| BC115747.1 | - | 1 | 41 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin3b| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130455.2 | NP_569722.1 | 3 | 1954 | ||
| GenBank mRNA | AF373861.1 | - | 1 | 703 | ||
| AF396649.1 | - | 3 | 1963 | |||
| AY062435.1 | - | 3 | 1954 | |||
| BC115747.1 | - | 1 | 41 | |||