siRNA Details
| Gene Symbol: Snx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130796.4 | NP_570614.2 | 2 | 1851 | ||
| GenBank mRNA | AF408408.1 | - | 2 | 1839 | ||
| AK052343.1 | - | 2 | 1812 | |||
| AK076119.1 | - | 2 | 679 | |||
| AK077890.1 | - | 2 | 1445 | |||
| BC040407.1 | - | 2 | 554 | |||
| BC063089.1 | - | 2 | 1851 | 6516923 | ||
| BC137801.1 | - | 2 | 1837 | 8860820 | ||
| BC145951.1 | - | 2 | 1837 | 40131254 | ||

Snx18
Genomic Map