siRNA Details
| Gene Symbol: Hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174998.4 | NP_778163.1 | 3 | 316 | ||
| GenBank mRNA | AB079893.1 | - | 2 | 108 | ||
| AK032591.1 | - | 3 | 316 | |||
| BC138631.1 | - | 2 | 151 | 8861651 | ||
| BC138632.1 | - | 2 | 151 | 8861652 | ||

Hpcal4
Genomic Map