siRNA Details
| Gene Symbol: Olfr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168503.1 | NP_001161975.1 | 3 | 849 | ||
| NM_130866.4 | NP_570936.1 | 4 | 1718 | |||
| GenBank mRNA | AF378854.1 | - | 1 | 503 | ||
| AK028467.1 | - | 4 | 1643 | |||
| AK036356.1 | - | 3 | 850 | |||
| BC120603.1 | - | 2 | 692 | 40129526 | ||
| BC120629.1 | - | 2 | 692 | 40129552 | ||

Olfr78
Genomic Map