siRNA Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170985.1 | NP_001164456.1 | 2 | 468 | ||
| NM_130867.3 | NP_570937.2 | 2 | 465 | |||
| GenBank mRNA | AF480411.1 | - | 1 | 103 | ||
| AK158381.1 | - | 2 | 422 | |||
| AY017368.1 | - | 1 | 103 | |||
| AY243095.1 | - | 1 | 103 | |||
| BC023765.1 | - | 2 | 281 | 5343720 | ||
| BY736743.1 | - | 2 | 251 | |||

Kirrel
Genomic Map