siRNA Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170985.1 | NP_001164456.1 | 12 | 1839 | ||
| NM_130867.3 | NP_570937.2 | 12 | 1836 | |||
| GenBank mRNA | AF480411.1 | - | 11 | 1474 | ||
| AK158381.1 | - | 12 | 1792 | |||
| AK209127.1 | - | 1 | 117 | |||
| AY243095.1 | - | 11 | 1474 | |||
| BC023765.1 | - | 12 | 1652 | 5343720 | ||

Kirrel
Genomic Map