siRNA Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170985.1 | NP_001164456.1 | 9, 10 | 1513 | ||
| NM_130867.3 | NP_570937.2 | 9, 10 | 1510 | |||
| GenBank mRNA | AF480411.1 | - | 8, 9 | 1148 | ||
| AK158381.1 | - | 9, 10 | 1467 | |||
| AY017368.1 | - | 8, 9 | 1148 | |||
| AY243095.1 | - | 8, 9 | 1148 | |||
| BC023765.1 | - | 9, 10 | 1326 | 5343720 | ||

Kirrel
Genomic Map