siRNA Details
Gene Symbol: Krtap19-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130870.1 | NP_570940.1 | 1 | 512 | ||
GenBank mRNA | AF345291.1 | - | 1 | 512 | ||
AK137740.1 | - | 1 | 208 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap19-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130870.1 | NP_570940.1 | 1 | 512 | ||
GenBank mRNA | AF345291.1 | - | 1 | 512 | ||
AK137740.1 | - | 1 | 208 |