siRNA Details
| Gene Symbol: Krtap19-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130870.1 | NP_570940.1 | 1 | 536 | ||
| GenBank mRNA | AF345291.1 | - | 1 | 536 | ||
| AK137740.1 | - | 1 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap19-2| Gene Symbol: Krtap19-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130870.1 | NP_570940.1 | 1 | 536 | ||
| GenBank mRNA | AF345291.1 | - | 1 | 536 | ||
| AK137740.1 | - | 1 | 232 | |||