siRNA Details
| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006193.2 | NP_006184.2 | 9 | 1855 | ||
| GenBank mRNA | AF043978.1 | - | 9 | 1853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAX4| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006193.2 | NP_006184.2 | 9 | 1855 | ||
| GenBank mRNA | AF043978.1 | - | 9 | 1853 | ||