siRNA Details
| Gene Symbol: Krtap19-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130876.3 | NP_570946.2 | 1 | 383 | ||
| GenBank mRNA | AA930779.1 | - | 1 | 75 | ||
| AF345292.1 | - | 1 | 340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap19-1| Gene Symbol: Krtap19-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130876.3 | NP_570946.2 | 1 | 383 | ||
| GenBank mRNA | AA930779.1 | - | 1 | 75 | ||
| AF345292.1 | - | 1 | 340 | |||