siRNA Details
| Gene Symbol: Peg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040611.1 | NP_001035701.1 | 2 | 621 | ||
| NM_130877.2 | NP_570947.2 | 2 | 621 | |||
| GenBank mRNA | AB091827.1 | - | 2 | 547 | ||
| AJ006464.1 | - | 2 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peg10| Gene Symbol: Peg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040611.1 | NP_001035701.1 | 2 | 621 | ||
| NM_130877.2 | NP_570947.2 | 2 | 621 | |||
| GenBank mRNA | AB091827.1 | - | 2 | 547 | ||
| AJ006464.1 | - | 2 | 531 | |||