siRNA Details
Gene Symbol: Otud7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130880.1 | NP_570950.1 | 10 | 1286 | ||
GenBank mRNA | AJ430384.1 | - | 10 | 1286 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Otud7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130880.1 | NP_570950.1 | 10 | 1286 | ||
GenBank mRNA | AJ430384.1 | - | 10 | 1286 |