siRNA Details
| Gene Symbol: Card14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130886.3 | NP_570956.1 | 18 | 2616 | ||
| GenBank mRNA | AF363457.1 | - | 18 | 2595 | ||
| AK028888.1 | - | 12 | 2001 | |||
| BC004692.1 | - | 14 | 1987 | |||
| BC029102.1 | - | 18 | 2616 | 3979883 | ||

Card14
Genomic Map