siRNA Details
| Gene Symbol: Rtn4ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130892.4 | NP_570962.2 | 4 | 972 | ||
| GenBank mRNA | AF336862.1 | - | 4 | 702 | ||
| AK050324.1 | - | 4 | 970 | |||
| AK085858.1 | - | 4 | 952 | |||
| AK088029.1 | - | 4 | 973 | |||
| AK204568.1 | - | 4 | 302 | |||
| BC024116.1 | - | 4 | 942 | 5100503 | ||

Rtn4ip1
Genomic Map