siRNA Details
| Gene Symbol: Scrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130893.3 | NP_570963.1 | 1, 2 | 532 | ||
| GenBank mRNA | BC057562.1 | - | 1, 2 | 305 | 6401572 | |
| BC089316.1 | - | 1, 2 | 316 | |||
| CB525529.1 | - | 1, 2 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scrt1| Gene Symbol: Scrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130893.3 | NP_570963.1 | 1, 2 | 532 | ||
| GenBank mRNA | BC057562.1 | - | 1, 2 | 305 | 6401572 | |
| BC089316.1 | - | 1, 2 | 316 | |||
| CB525529.1 | - | 1, 2 | 532 | |||