siRNA Details
| Gene Symbol: Znrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168621.1 | NP_001162092.1 | 2 | 975 | ||
| NM_001168623.1 | NP_001162094.1 | 2 | 174 | |||
| NM_133206.3 | NP_573469.1 | 2 | 975 | |||
| GenBank mRNA | AF378525.1 | - | 2 | 680 | ||
| AK076756.1 | - | 2 | 122 | |||
| AK078819.1 | - | 2 | 201 | |||
| AK079338.1 | - | 2 | 975 | |||
| BC006765.1 | - | 2 | 575 | 3710377 | ||
| BC086770.1 | - | 2 | 773 | 5720382 | ||

Znrf1
Genomic Map