siRNA Details
| Gene Symbol: Smco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133214.2 | NP_573477.2 | 3 | 901 | ||
| GenBank mRNA | AF197136.1 | - | 3 | 901 | ||
| AK146267.1 | - | 3 | 944 | |||
| BC017612.1 | - | 3 | 779 | 4911985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smco4| Gene Symbol: Smco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133214.2 | NP_573477.2 | 3 | 901 | ||
| GenBank mRNA | AF197136.1 | - | 3 | 901 | ||
| AK146267.1 | - | 3 | 944 | |||
| BC017612.1 | - | 3 | 779 | 4911985 | ||