siRNA Details
| Gene Symbol: Mtmr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133215.1 | NP_573478.1 | 14 | 1645 | ||
| GenBank mRNA | AF262986.1 | - | 14 | 1645 | ||
| AK146641.1 | - | 14 | 1662 | |||
| AK186095.1 | - | 2 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr4| Gene Symbol: Mtmr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133215.1 | NP_573478.1 | 14 | 1645 | ||
| GenBank mRNA | AF262986.1 | - | 14 | 1645 | ||
| AK146641.1 | - | 14 | 1662 | |||
| AK186095.1 | - | 2 | 214 | |||