siRNA Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133216.3 | NP_573479.3 | 18 | 1796 | ||
| GenBank mRNA | AF363970.1 | - | 18 | 1736 | ||
| AK052094.1 | - | 10 | 938 | |||
| AK080423.1 | - | 7 | 544 | |||
| AK146033.1 | - | 18 | 1776 | |||
| AK149676.1 | - | 18 | 1735 | |||
| AK158597.1 | - | 16 | 1754 | |||
| AK166715.1 | - | 18 | 1696 | |||
| AK169024.1 | - | 17 | 1622 | |||
| BC021534.1 | - | 9 | 731 | |||
| BC065174.1 | - | 18 | 1684 | 6415314 | ||

Xpnpep1
Genomic Map