siRNA Details
| Gene Symbol: Zfp704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133218.2 | NP_573481.1 | 2 | 848 | ||
| GenBank mRNA | AF292939.1 | - | 2 | 848 | ||
| CJ177621.1 | - | 2 | 289 | |||
| DT908776.1 | - | 4 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp704| Gene Symbol: Zfp704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133218.2 | NP_573481.1 | 2 | 848 | ||
| GenBank mRNA | AF292939.1 | - | 2 | 848 | ||
| CJ177621.1 | - | 2 | 289 | |||
| DT908776.1 | - | 4 | 212 | |||