siRNA Details
| Gene Symbol: Zfp704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133218.2 | NP_573481.1 | 9 | 1994 | ||
| GenBank mRNA | AF292939.1 | - | 9 | 1994 | ||
| AK078492.1 | - | 5 | 542 | |||
| AK164533.1 | - | 2 | 1290 | |||
| BC052936.1 | - | 6 | 728 | |||

Zfp704
Genomic Map