siRNA Details
| Gene Symbol: Nup155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133227.3 | NP_573490.3 | 18 | 2141 | ||
| GenBank mRNA | AF322375.1 | - | 18 | 2127 | ||
| AK036575.1 | - | 18 | 2109 | |||
| AK086426.1 | - | 18 | 2115 | |||
| AK129217.1 | - | 18 | 2104 | |||
| AK145717.1 | - | 18 | 2119 | |||
| BC050919.1 | - | 18 | 2094 | 6406277 | ||

Nup155
Genomic Map