siRNA Details
| Gene Symbol: Zfp87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133228.3 | NP_573491.2 | 4 | 1131 | ||
| GenBank mRNA | AF242377.1 | - | 4 | 1034 | ||
| AK028465.1 | - | 1 | 687 | |||
| AK030724.1 | - | 4 | 1079 | |||
| AK049711.1 | - | 4 | 1051 | |||
| BC031988.1 | - | 4 | 1034 | 4017149 | ||

Zfp87
Genomic Map