siRNA Details
| Gene Symbol: Ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133229.2 | NP_573492.2 | 3, 4 | 331 | ||
| GenBank mRNA | AB063284.1 | - | 3, 4 | 333 | ||
| AK041355.1 | - | 3, 4 | 321 | |||
| AK172430.1 | - | 3, 4 | 324 | |||
| BC060291.1 | - | 3, 4 | 289 | 30135244 | ||
| CF170896.1 | - | 3, 4 | 331 | |||

Ripply3
Genomic Map