siRNA Details
| Gene Symbol: Bbc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133234.2 | NP_573497.1 | 4 | 1747 | ||
| GenBank mRNA | AF332560.1 | - | 4 | 1691 | ||
| BC044782.2 | - | 4 | 1712 | |||
| BC060370.1 | - | 4 | 1677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbc3| Gene Symbol: Bbc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133234.2 | NP_573497.1 | 4 | 1747 | ||
| GenBank mRNA | AF332560.1 | - | 4 | 1691 | ||
| BC044782.2 | - | 4 | 1712 | |||
| BC060370.1 | - | 4 | 1677 | |||