siRNA Details
| Gene Symbol: Glcci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286728.1 | NP_001273657.1 | 6 | 615 | ||
| NM_001286729.1 | NP_001273658.1 | 6 | 612 | |||
| NM_133236.3 | NP_573499.2 | 6 | 1288 | |||
| GenBank mRNA | AK029524.1 | - | 6 | 614 | ||
| AY098636.1 | - | 6 | 1334 | |||
| BC050802.1 | - | 6 | 653 | 6771009 | ||
| DV652660.1 | - | 2 | 187 | |||

Glcci1
Genomic Map