siRNA Details
| Gene Symbol: Glcci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286728.1 | NP_001273657.1 | 2 | 104 | ||
| NM_001286729.1 | NP_001273658.1 | 2 | 104 | |||
| NM_133236.3 | NP_573499.2 | 2 | 780 | |||
| GenBank mRNA | AK029524.1 | - | 2 | 106 | ||
| AY098636.1 | - | 2 | 826 | |||
| BC050802.1 | - | 2 | 142 | 6771009 | ||
| CA561411.1 | - | 2 | 160 | |||
| CN720421.1 | - | 2 | 137 | |||

Glcci1
Genomic Map